Learn More
MSC Vermont Gage Pin Gage, Class ZZ, 4.26 mm, Minus Tolerance, Steel
Catalog No. 5000528908
Encompass
This Class ZZ Pin Gage is a precision measuring tool used for setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection. It is designed for accuracy and durability in various industrial applications.
  • Class ZZ tolerance
  • 4.26 mm diameter
  • Minus tolerance direction
  • Made of steel
  • Reversible style
  • Metric system of measurement
  • 0.00508 mm tolerance
Catalog No. 50-005-28908 Supplier MSC Supplier No. 40431710
This Class ZZ Pin Gage is a precision measuring tool used for setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection. It is designed for accuracy and durability in various industrial applications.
  • Class ZZ tolerance
  • 4.26 mm diameter
  • Minus tolerance direction
  • Made of steel
  • Reversible style
  • Metric system of measurement
  • 0.00508 mm tolerance