Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Go, 7.77 mm, Tool Steel
Catalog No. 5000527331
Encompass
This Vermont Gage pin gage is a precision tool designed for 'Go' inspection and for accurately setting various measuring devices, including micrometers and verniers. Manufactured from durable tool steel, this Class ZZ gage ensures reliability and accuracy for critical measurement tasks.
  • Class ZZ designation for high precision
  • Go (Plus) type
  • Constructed from tool steel with a Rockwell hardness of Rc 60/65
  • Features a steel finish with 10 microfinish or better
  • Includes a certificate of accuracy with NIST traceability
  • Overall length of 2 inches
Catalog No. 50-005-27331 Supplier MSC Supplier No. 68098821
This Vermont Gage pin gage is a precision tool designed for 'Go' inspection and for accurately setting various measuring devices, including micrometers and verniers. Manufactured from durable tool steel, this Class ZZ gage ensures reliability and accuracy for critical measurement tasks.
  • Class ZZ designation for high precision
  • Go (Plus) type
  • Constructed from tool steel with a Rockwell hardness of Rc 60/65
  • Features a steel finish with 10 microfinish or better
  • Includes a certificate of accuracy with NIST traceability
  • Overall length of 2 inches