Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Go (Plus), 0.342 in Diameter, +.0002/-.0000 Tolerance
Catalog No. 5000537574
Encompass
This pin gage is designed for precise measurement of hole sizes. Constructed from durable tool steel with a fine finish, it ensures accuracy and longevity. It comes with a certificate of accuracy with NIST traceability for reliable calibration.
  • Constructed from tool steel with a hardness of Rc 60/65 for durability and precision.
  • Features a steel - 10 microfinish or better, contributing to accuracy and longevity.
  • Maintains a tight tolerance of +.0002/-.0000, suitable for precise measurements of hole sizes.
  • Includes a certificate of accuracy with NIST traceability, offering assurance of calibration standards.
  • "Go (Plus)" designation indicates its function in quality control for checking minimum acceptable sizes.
Catalog No. 50-005-37574 Supplier MSC Supplier No. 88934203
This pin gage is designed for precise measurement of hole sizes. Constructed from durable tool steel with a fine finish, it ensures accuracy and longevity. It comes with a certificate of accuracy with NIST traceability for reliable calibration.
  • Constructed from tool steel with a hardness of Rc 60/65 for durability and precision.
  • Features a steel - 10 microfinish or better, contributing to accuracy and longevity.
  • Maintains a tight tolerance of +.0002/-.0000, suitable for precise measurements of hole sizes.
  • Includes a certificate of accuracy with NIST traceability, offering assurance of calibration standards.
  • "Go (Plus)" designation indicates its function in quality control for checking minimum acceptable sizes.