Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Go, Steel, 1.73 mm
Catalog No. 5000512492
Encompass
This Class ZZ pin gage is a "Go" plug gage made of tool steel, with a diameter of 1.73 mm. It is designed for "Go" inspection and can also be used to set micrometers, verniers, and other measuring devices.
  • For go inspection.
  • Used to set micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance.
  • 1.73 mm diameter.
  • Tool steel material.
  • Reversible style.
  • Metric system of measurement.
  • Plus direction of tolerance.
Catalog No. 50-005-12492 Supplier MSC Supplier No. 68092782
This Class ZZ pin gage is a "Go" plug gage made of tool steel, with a diameter of 1.73 mm. It is designed for "Go" inspection and can also be used to set micrometers, verniers, and other measuring devices.
  • For go inspection.
  • Used to set micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance.
  • 1.73 mm diameter.
  • Tool steel material.
  • Reversible style.
  • Metric system of measurement.
  • Plus direction of tolerance.