Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.0130 in Diameter, 2 in OAL
Catalog No. 5000533813
Encompass
This pin gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Constructed from steel with a black oxide finish, it features a 0.0130 in diameter and a 2 in overall length, with a tolerance of 0.000200 in.
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-33813 Supplier MSC Supplier No. 37242047
This pin gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Constructed from steel with a black oxide finish, it features a 0.0130 in diameter and a 2 in overall length, with a tolerance of 0.000200 in.
  • Includes certificate of accuracy with NIST traceability