Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.0350 in, Steel
Catalog No. 5000530134
Encompass
This Class ZZ Minus pin gage is designed for precise measurement and inspection applications. Made from durable steel with a black oxide finish, it features a 0.0350 inch diameter and a 2 inch overall length, providing reliable accuracy for various gaging tasks.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection and gaging applications.
  • Constructed from steel with a black oxide coating for durability.
  • Provides a precise 0.0350 inch diameter.
  • Features a 2 inch overall length.
  • Includes a certificate of accuracy with NIST traceability.
  • Maintains a tolerance of 0.00020 "
  • Hardness of RC 60/65.
  • Surface finish of 10 microinch or better.
  • Meets ISO 17025 standards.
Catalog No. 50-005-30134 Supplier MSC Supplier No. 78651429
This Class ZZ Minus pin gage is designed for precise measurement and inspection applications. Made from durable steel with a black oxide finish, it features a 0.0350 inch diameter and a 2 inch overall length, providing reliable accuracy for various gaging tasks.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection and gaging applications.
  • Constructed from steel with a black oxide coating for durability.
  • Provides a precise 0.0350 inch diameter.
  • Features a 2 inch overall length.
  • Includes a certificate of accuracy with NIST traceability.
  • Maintains a tolerance of 0.00020 "
  • Hardness of RC 60/65.
  • Surface finish of 10 microinch or better.
  • Meets ISO 17025 standards.