Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.1710 in Diameter, 2 in OAL
Catalog No. 5000529984
Encompass
This pin gage is a Class ZZ No-Go type, designed for precise measurement of hole sizes. It features a steel construction with a black oxide finish, providing durability and accuracy for various applications. The gage is manufactured to a tolerance of 0.00020 inch and includes a certificate of accuracy with NIST traceability.
  • Steel construction
  • Tolerance of 0.00020 in
  • Measures hole sizes within +0.0000/-0.0002 in
  • Includes certificate of accuracy with NIST traceability
  • Overall length of 2 in
  • Class ZZ no-go pin gauge
  • Black oxide finish
  • Surface finish of 10 microinch or better
  • Hardness of RC 60/65
  • Meets ISO 17025 standards
Catalog No. 50-005-29984 Supplier MSC Supplier No. 06016091
This pin gage is a Class ZZ No-Go type, designed for precise measurement of hole sizes. It features a steel construction with a black oxide finish, providing durability and accuracy for various applications. The gage is manufactured to a tolerance of 0.00020 inch and includes a certificate of accuracy with NIST traceability.
  • Steel construction
  • Tolerance of 0.00020 in
  • Measures hole sizes within +0.0000/-0.0002 in
  • Includes certificate of accuracy with NIST traceability
  • Overall length of 2 in
  • Class ZZ no-go pin gauge
  • Black oxide finish
  • Surface finish of 10 microinch or better
  • Hardness of RC 60/65
  • Meets ISO 17025 standards