Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.1750 in Diameter, 2 in Overall Length, Steel
Catalog No. 5000529146
Encompass
This Class ZZ No-Go Pin Gauge is designed for quick and accurate measurements of hole diameters, distances between holes, slot widths, and locations. It can also be used for Go/NoGo inspection and to set other measuring devices. Made of steel with a black oxide finish, it has a diameter of 0.1750 inches and an overall length of 2 inches, with a tolerance of 0.00020 inches.
  • Made of steel with a black oxide finish
  • Diameter of 0.1750 in and 2 in overall length
  • Class ZZ tolerance of 0.00020 in
  • Designed for Go/NoGo inspection and precise hole size measurement
  • Surface finish of 10 microinch or better
  • Hardness of RC 60/65
  • Includes a certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards
  • Cylindrical shape
Catalog No. 50-005-29146 Supplier MSC Supplier No. 06016125
This Class ZZ No-Go Pin Gauge is designed for quick and accurate measurements of hole diameters, distances between holes, slot widths, and locations. It can also be used for Go/NoGo inspection and to set other measuring devices. Made of steel with a black oxide finish, it has a diameter of 0.1750 inches and an overall length of 2 inches, with a tolerance of 0.00020 inches.
  • Made of steel with a black oxide finish
  • Diameter of 0.1750 in and 2 in overall length
  • Class ZZ tolerance of 0.00020 in
  • Designed for Go/NoGo inspection and precise hole size measurement
  • Surface finish of 10 microinch or better
  • Hardness of RC 60/65
  • Includes a certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards
  • Cylindrical shape