Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.176 in, 2 in Length
Catalog No. 5000530897
Encompass
This Class ZZ Minus Pin Gage is designed for precise measurement of hole sizes, slot widths, and locations. It provides an economical solution for 'NoGo' gaging applications, ensuring extended wear life for consistent performance.
  • Made from durable tool steel with 60/65 Rc hardness.
  • Features a Class ZZ tolerance of 0.0002 "
  • Measures 0.1760 inch in diameter and is 2 inches long.
  • Offers a 10 microinch or better surface finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Meets or exceeds ISO 17025 standards.
Catalog No. 50-005-30897 Supplier MSC Supplier No. 78373727
This Class ZZ Minus Pin Gage is designed for precise measurement of hole sizes, slot widths, and locations. It provides an economical solution for 'NoGo' gaging applications, ensuring extended wear life for consistent performance.
  • Made from durable tool steel with 60/65 Rc hardness.
  • Features a Class ZZ tolerance of 0.0002 "
  • Measures 0.1760 inch in diameter and is 2 inches long.
  • Offers a 10 microinch or better surface finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Meets or exceeds ISO 17025 standards.