Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.2825 in, 2 in
Catalog No. 5000515200
Encompass
This precision gage is a Class ZZ Minus plug and pin gage, manufactured from steel. It features a 0.2825 inch diameter and a 2 inch overall length with a 10 microfinish.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for NoGo inspection.
  • Can be used for setting micrometers, verniers, and other measuring devices.
Catalog No. 50-005-15200 Supplier MSC Supplier No. 89028252
This precision gage is a Class ZZ Minus plug and pin gage, manufactured from steel. It features a 0.2825 inch diameter and a 2 inch overall length with a 10 microfinish.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for NoGo inspection.
  • Can be used for setting micrometers, verniers, and other measuring devices.