Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.505 inch Diameter, 2 inch Length
Catalog No. 5000515269
Encompass
This general purpose pin gage is designed for quick and accurate measurements, making it suitable for tasks such as measuring hole sizes, calculating distances between holes, gaging slot widths, checking locations, and Go/NoGo gaging. Constructed from durable tool steel, it offers enhanced wear life and can be used to update existing gage sets.
  • Economical and versatile for various precision measurement applications.
  • Provides quick and accurate measurements.
  • Offers extended wear life.
  • Ideal for updating and maintaining compliance of existing gage sets.
  • Individually packaged.
  • Includes a Certificate of Accuracy.
  • NIST traceable for reliability.
  • Features a durable 2 inch length.
  • Achieves a smooth 10 microinch finish or better.
  • Marked with size and direction of tolerance on gages over 0.0605 inch for easy identification.
Catalog No. 50-005-15269 Supplier MSC Supplier No. 89050504
This general purpose pin gage is designed for quick and accurate measurements, making it suitable for tasks such as measuring hole sizes, calculating distances between holes, gaging slot widths, checking locations, and Go/NoGo gaging. Constructed from durable tool steel, it offers enhanced wear life and can be used to update existing gage sets.
  • Economical and versatile for various precision measurement applications.
  • Provides quick and accurate measurements.
  • Offers extended wear life.
  • Ideal for updating and maintaining compliance of existing gage sets.
  • Individually packaged.
  • Includes a Certificate of Accuracy.
  • NIST traceable for reliability.
  • Features a durable 2 inch length.
  • Achieves a smooth 10 microinch finish or better.
  • Marked with size and direction of tolerance on gages over 0.0605 inch for easy identification.