Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.6180 in Diameter, 2 in OAL
Catalog No. 5000531651
Encompass
This Vermont Gage Class ZZ Minus Pin Gage, made from durable tool steel, offers precise measurement for inspection applications. With a 0.6180 inch diameter, 2 inch overall length, and a tight tolerance of +.0000/-.0002, it's ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Includes a Certificate of Accuracy with NIST traceability.
  • Constructed from high-quality tool steel with a 10 microfinish.
  • Features a precise 0.6180 inch diameter and 2 inch overall length.
  • Maintains a tight tolerance of +.0000/-.0002.
  • Suitable for setting micrometers, NoGo inspection, and use with various measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-31651 Supplier MSC Supplier No. 89061808
This Vermont Gage Class ZZ Minus Pin Gage, made from durable tool steel, offers precise measurement for inspection applications. With a 0.6180 inch diameter, 2 inch overall length, and a tight tolerance of +.0000/-.0002, it's ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Includes a Certificate of Accuracy with NIST traceability.
  • Constructed from high-quality tool steel with a 10 microfinish.
  • Features a precise 0.6180 inch diameter and 2 inch overall length.
  • Maintains a tight tolerance of +.0000/-.0002.
  • Suitable for setting micrometers, NoGo inspection, and use with various measuring devices.
  • Includes a certificate of accuracy with NIST traceability.