Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.8580 in, Tool Steel
Catalog No. 5000531184
Encompass
This pin gage is designed for precise measurement of hole sizes. Constructed from durable tool steel with a Rockwell C hardness of 60-65, it features a 0.8580 inch diameter and a tight tolerance of +.0000/-.0002. The gage has an overall length of 2 inches and is a Class ZZ minus type, suitable for NoGo applications.
  • Tool steel construction with Rc 60/65 hardness
  • Steel finish with 10 microfinish or better
  • 0.8580 inch diameter
  • Tolerance of +.0000/-.0002
  • 2 inch overall length
  • Measures hole sizes precisely
  • Includes certificate of accuracy with NIST traceability
  • NoGo (minus) type
  • Class ZZ
Catalog No. 50-005-31184 Supplier MSC Supplier No. 89085807
This pin gage is designed for precise measurement of hole sizes. Constructed from durable tool steel with a Rockwell C hardness of 60-65, it features a 0.8580 inch diameter and a tight tolerance of +.0000/-.0002. The gage has an overall length of 2 inches and is a Class ZZ minus type, suitable for NoGo applications.
  • Tool steel construction with Rc 60/65 hardness
  • Steel finish with 10 microfinish or better
  • 0.8580 inch diameter
  • Tolerance of +.0000/-.0002
  • 2 inch overall length
  • Measures hole sizes precisely
  • Includes certificate of accuracy with NIST traceability
  • NoGo (minus) type
  • Class ZZ