Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 1.76 mm, Tool Steel
Catalog No. 5000532806
Encompass
This pin gage is designed for precise measurement of hole sizes. It features a 1.76 mm diameter and is constructed from durable tool steel. Ideal for quality control and inspection, this gage ensures accurate measurements and comes with a Certificate of Accuracy that provides NIST traceability.
  • Made from tool steel with Rc 60/65 hardness.
  • Measures hole sizes with a tolerance of +.000/-.005 in.
  • Includes certificate of accuracy with NIST traceability.
  • Features an overall length of 2 in.
  • Boasts a steel finish of 10 microfinish or better.
  • Designed as a NoGo (Minus) type gage.
  • Class ZZ for specific measurement requirements.
Catalog No. 50-005-32806 Supplier MSC Supplier No. 40429219
This pin gage is designed for precise measurement of hole sizes. It features a 1.76 mm diameter and is constructed from durable tool steel. Ideal for quality control and inspection, this gage ensures accurate measurements and comes with a Certificate of Accuracy that provides NIST traceability.
  • Made from tool steel with Rc 60/65 hardness.
  • Measures hole sizes with a tolerance of +.000/-.005 in.
  • Includes certificate of accuracy with NIST traceability.
  • Features an overall length of 2 in.
  • Boasts a steel finish of 10 microfinish or better.
  • Designed as a NoGo (Minus) type gage.
  • Class ZZ for specific measurement requirements.