Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 10.16 mm, 2 in, Tool Steel
Catalog No. 5000514431
Encompass
This pin gage is a Class ZZ Minus instrument, precisely crafted from tool steel with a 10.16 mm diameter and a 2 in overall length. It is primarily used for accurately measuring hole sizes and can also serve to set and calibrate various measuring devices.
  • Measures hole sizes within a +.000/-.005" tolerance
  • Suitable for no-go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from tool steel, Rc 60/65
  • Features a 10 microfinish or better
Catalog No. 50-005-14431 Supplier MSC Supplier No. 40437618
This pin gage is a Class ZZ Minus instrument, precisely crafted from tool steel with a 10.16 mm diameter and a 2 in overall length. It is primarily used for accurately measuring hole sizes and can also serve to set and calibrate various measuring devices.
  • Measures hole sizes within a +.000/-.005" tolerance
  • Suitable for no-go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from tool steel, Rc 60/65
  • Features a 10 microfinish or better