Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Minus, 11.14 mm, Tool Steel
Catalog No. 5000527897
Encompass
This precision measuring tool is crafted from durable tool steel with a hardness rating of Rc 60/65. It features a steel - 10 microfinish or better for enhanced accuracy and longevity. With a diameter of 11.14 mm and an overall length of 2 inches, this NoGo (Minus) Class ZZ pin gage is designed to precisely measure hole sizes within a tight tolerance. Each gage includes a certificate of accuracy with NIST traceability, ensuring reliable and verifiable measurements.
  • Manufactured from durable tool steel with a hardness of Rc 60/65.
  • Features a steel - 10 microfinish or better for enhanced accuracy.
  • Overall length of 2 inches.
  • Designed as a NoGo (minus) Class ZZ pin gage.
  • Measures hole sizes with a tolerance of +.000/-.005 inches.
  • Includes a certificate of accuracy with NIST traceability.
  • Round within .0001 inch or 0.0025 mm.
  • Made in the United States.
Catalog No. 50-005-27897 Supplier MSC Supplier No. 40438590
This precision measuring tool is crafted from durable tool steel with a hardness rating of Rc 60/65. It features a steel - 10 microfinish or better for enhanced accuracy and longevity. With a diameter of 11.14 mm and an overall length of 2 inches, this NoGo (Minus) Class ZZ pin gage is designed to precisely measure hole sizes within a tight tolerance. Each gage includes a certificate of accuracy with NIST traceability, ensuring reliable and verifiable measurements.
  • Manufactured from durable tool steel with a hardness of Rc 60/65.
  • Features a steel - 10 microfinish or better for enhanced accuracy.
  • Overall length of 2 inches.
  • Designed as a NoGo (minus) Class ZZ pin gage.
  • Measures hole sizes with a tolerance of +.000/-.005 inches.
  • Includes a certificate of accuracy with NIST traceability.
  • Round within .0001 inch or 0.0025 mm.
  • Made in the United States.