Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 13.93 mm, Tool Steel
Catalog No. 5000529191
Encompass
This Class ZZ Minus pin gage is engineered for precise no-go inspection, ideal for verifying hole sizes. Constructed from hardened tool steel with a superior finish, it ensures consistent and accurate measurements.
  • Made from tool steel, Rc 60/65.
  • Features a 0.005 mm tolerance.
  • Surface finish is 10 microinch or better.
  • Overall length measures 2 in.
  • Includes certificate of accuracy with NIST traceability.
Catalog No. 50-005-29191 Supplier MSC Supplier No. 40441388
This Class ZZ Minus pin gage is engineered for precise no-go inspection, ideal for verifying hole sizes. Constructed from hardened tool steel with a superior finish, it ensures consistent and accurate measurements.
  • Made from tool steel, Rc 60/65.
  • Features a 0.005 mm tolerance.
  • Surface finish is 10 microinch or better.
  • Overall length measures 2 in.
  • Includes certificate of accuracy with NIST traceability.