Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 15.70 mm, 2 in
Catalog No. 5000512056
Encompass
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection and calibration tasks. This steel gage features a 15.70 mm diameter and 2-inch overall length, with a 10 Micro finish for precise measurements. It is commonly used for setting micrometers, performing NoGo inspections, and with other measuring devices.
  • Designed for accurate inspection and calibration.
  • Constructed from durable steel for longevity.
  • Features a 15.70 mm diameter for specific measurements.
  • Comes with a 2-inch overall length.
  • Provides a 10 Micro finish for high precision.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers and NoGo inspections.
  • Compatible with verniers and other measuring devices.
  • Has a tolerance of 0.00508 mm.
Catalog No. 50-005-12056 Supplier MSC Supplier No. 40443152
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection and calibration tasks. This steel gage features a 15.70 mm diameter and 2-inch overall length, with a 10 Micro finish for precise measurements. It is commonly used for setting micrometers, performing NoGo inspections, and with other measuring devices.
  • Designed for accurate inspection and calibration.
  • Constructed from durable steel for longevity.
  • Features a 15.70 mm diameter for specific measurements.
  • Comes with a 2-inch overall length.
  • Provides a 10 Micro finish for high precision.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers and NoGo inspections.
  • Compatible with verniers and other measuring devices.
  • Has a tolerance of 0.00508 mm.