Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 16.40 mm, Steel
Catalog No. 5000514158
Encompass
Class ZZ Minus pin gages are economical, general purpose gages for quick, accurate measurements. They are used for measuring hole sizes, calculating distances, gaging slot widths, checking locations, and Go/NoGo gaging. This pin gage is tool steel, 16.40 mm diameter, 2 in length, with a 10 microinch finish, offering extended wear life.
  • Economical general purpose pin gages
  • Quick and accurate measurements
  • Measures hole sizes, distances, slot widths
  • Checks locations and Go/NoGo gaging
  • Extended wear life
  • Class ZZ tolerance (.005 mm)
  • NoGo (minus) tolerance
  • Durable tool steel (60/65 Rc)
  • 2 in overall length
  • Round within .0025 mm
  • 10 microinch finish or better
  • NIST traceable, with certificate of accuracy
Catalog No. 50-005-14158 Supplier MSC Supplier No. 40443855
Class ZZ Minus pin gages are economical, general purpose gages for quick, accurate measurements. They are used for measuring hole sizes, calculating distances, gaging slot widths, checking locations, and Go/NoGo gaging. This pin gage is tool steel, 16.40 mm diameter, 2 in length, with a 10 microinch finish, offering extended wear life.
  • Economical general purpose pin gages
  • Quick and accurate measurements
  • Measures hole sizes, distances, slot widths
  • Checks locations and Go/NoGo gaging
  • Extended wear life
  • Class ZZ tolerance (.005 mm)
  • NoGo (minus) tolerance
  • Durable tool steel (60/65 Rc)
  • 2 in overall length
  • Round within .0025 mm
  • 10 microinch finish or better
  • NIST traceable, with certificate of accuracy