Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 16.49 mm Diameter, 2 in Length
Catalog No. 5000514014
Encompass
This precision pin gage is designed for accurate measurement and calibration in various laboratory and industrial settings. It is ideal for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices, ensuring high precision in your work. Made from durable steel, this gage offers reliable performance.
  • Provides precise measurements for calibration tasks.
  • Suitable for NoGo inspection to verify tolerances.
  • Compatible with verniers and a range of other measuring devices.
  • Includes a certificate of accuracy with NIST traceability for assured quality.
  • Constructed from durable steel for long-lasting use.
  • Features a 10 micro coating/finish for enhanced performance.
Catalog No. 50-005-14014 Supplier MSC Supplier No. 40443947
This precision pin gage is designed for accurate measurement and calibration in various laboratory and industrial settings. It is ideal for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices, ensuring high precision in your work. Made from durable steel, this gage offers reliable performance.
  • Provides precise measurements for calibration tasks.
  • Suitable for NoGo inspection to verify tolerances.
  • Compatible with verniers and a range of other measuring devices.
  • Includes a certificate of accuracy with NIST traceability for assured quality.
  • Constructed from durable steel for long-lasting use.
  • Features a 10 micro coating/finish for enhanced performance.