Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 21.54 mm Diameter, Steel
Catalog No. 5000532171
Encompass
Class ZZ Minus pin gages are economical, general-purpose tools designed for quick and accurate measurements. Made of steel with a 10 microinch finish and a 2 inch overall length, they are suitable for no-go inspection and setting various measuring devices.
  • Economical general-purpose pin gage
  • Quick and accurate measurements
  • Class ZZ tolerance (0.00508 mm)
  • NIST traceability
  • Certificate of accuracy included
  • Material: Steel
  • 10 microinch finish or better
  • Overall length: 2 inches
  • Suitable for no-go inspection
  • Can be used to set micrometers, verniers, and other measuring devices
Catalog No. 50-005-32171 Supplier MSC Supplier No. 40448995
Class ZZ Minus pin gages are economical, general-purpose tools designed for quick and accurate measurements. Made of steel with a 10 microinch finish and a 2 inch overall length, they are suitable for no-go inspection and setting various measuring devices.
  • Economical general-purpose pin gage
  • Quick and accurate measurements
  • Class ZZ tolerance (0.00508 mm)
  • NIST traceability
  • Certificate of accuracy included
  • Material: Steel
  • 10 microinch finish or better
  • Overall length: 2 inches
  • Suitable for no-go inspection
  • Can be used to set micrometers, verniers, and other measuring devices