Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Minus, Steel, 6.13 mm
Catalog No. 5000516464
Encompass
This pin gage is a Class ZZ Minus gage with a 6.13 mm diameter, designed for quick and accurate measurements in various applications. Made from steel and featuring a 10 microinch finish, it is ideal for setting micrometers, Go/NoGo inspection, and verifying other measuring devices. Each gage is 2 inches long and comes with NIST traceability.
  • Economical, general-purpose pin gage
  • Quick and accurate measurements
  • Class ZZ tolerance for precision
  • Minus (NoGo) tolerance type
  • Durable steel construction
  • 6.13 mm diameter for specific gaging tasks
  • Two inches long for convenient handling
  • Smooth 10 microinch finish or better
  • NIST traceable for quality assurance
  • Includes a certificate of accuracy
Catalog No. 50-005-16464 Supplier MSC Supplier No. 40433583
This pin gage is a Class ZZ Minus gage with a 6.13 mm diameter, designed for quick and accurate measurements in various applications. Made from steel and featuring a 10 microinch finish, it is ideal for setting micrometers, Go/NoGo inspection, and verifying other measuring devices. Each gage is 2 inches long and comes with NIST traceability.
  • Economical, general-purpose pin gage
  • Quick and accurate measurements
  • Class ZZ tolerance for precision
  • Minus (NoGo) tolerance type
  • Durable steel construction
  • 6.13 mm diameter for specific gaging tasks
  • Two inches long for convenient handling
  • Smooth 10 microinch finish or better
  • NIST traceable for quality assurance
  • Includes a certificate of accuracy