Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Minus Tolerance, 5.45 mm, Tool Steel
Catalog No. 5000514584
Encompass
These economical general-purpose pin gages are designed for quick and accurate measurements. They can be used to measure hole sizes, calculate distances between holes, gage slot widths, check locations, and for Go/NoGo gaging. The gages offer added wear life compared to closer tolerance gages.
  • Class ZZ (.0002 in or .005 mm tolerance)
  • Available as Go (Plus) or NoGo (Minus)
  • NIST traceable
  • Available in .0005 in or .02 mm sizes
  • Increments of .001 in or .02 mm for sets
  • Gages over .0605 in are marked with size and direction of tolerance
  • Certificate of accuracy included
  • Individually packaged replacement pins include a certificate of accuracy
  • Allows quick and accurate measurements
  • Provides added wear life
Catalog No. 50-005-14584 Supplier MSC Supplier No. 40432908
These economical general-purpose pin gages are designed for quick and accurate measurements. They can be used to measure hole sizes, calculate distances between holes, gage slot widths, check locations, and for Go/NoGo gaging. The gages offer added wear life compared to closer tolerance gages.
  • Class ZZ (.0002 in or .005 mm tolerance)
  • Available as Go (Plus) or NoGo (Minus)
  • NIST traceable
  • Available in .0005 in or .02 mm sizes
  • Increments of .001 in or .02 mm for sets
  • Gages over .0605 in are marked with size and direction of tolerance
  • Certificate of accuracy included
  • Individually packaged replacement pins include a certificate of accuracy
  • Allows quick and accurate measurements
  • Provides added wear life