Vermont Gage pin gages provide quick and accurate measurements for various applications. They are designed to measure hole sizes, calculate distances between holes, and gage slot widths and locations. Suitable for Go/NoGo inspection and for setting micrometers and other measuring devices, this Class ZZ Minus Pin Gage has a 5.54 mm diameter and 2 inch overall length. It is constructed from tool steel with a 60/65 Rc hardness and features a 10 microinch finish or better.
- Class ZZ tolerance of 0.005 mm
- Minus tolerance design for NoGo gaging
- 2 inch overall length
- 10 microinch finish or better
- Round within 0.0025 mm
- Gages over 0.0605 in are marked with size and direction of tolerance
- Includes a Certificate of Accuracy with NIST Traceability
- Constructed from tool steel with 60/65 Rc hardness