Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, Tool Steel, 0.4260 in, 2 in
Catalog No. 5000516283
Encompass
This Class ZZ Minus pin gage is crafted from durable tool steel with a precise 0.4260 in diameter and 2 in overall length. Designed for accurate measurement, it is ideal for verifying hole sizes, gaging slot widths, and setting measuring devices in various industrial and laboratory applications. Its Class ZZ tolerance ensures reliability for Go/NoGo inspection tasks.
  • Made from robust tool steel (Rc 60/65) for durability.
  • Features a 10 microinch finish or better.
  • Provides a Class ZZ tolerance of +.0000/-.0002 in.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Suitable for Go/NoGo inspection and measuring hole sizes.
  • Includes a certificate of accuracy with NIST traceability.
  • Offers economical wear life for various gaging applications.
Catalog No. 50-005-16283 Supplier MSC Supplier No. 89042600
This Class ZZ Minus pin gage is crafted from durable tool steel with a precise 0.4260 in diameter and 2 in overall length. Designed for accurate measurement, it is ideal for verifying hole sizes, gaging slot widths, and setting measuring devices in various industrial and laboratory applications. Its Class ZZ tolerance ensures reliability for Go/NoGo inspection tasks.
  • Made from robust tool steel (Rc 60/65) for durability.
  • Features a 10 microinch finish or better.
  • Provides a Class ZZ tolerance of +.0000/-.0002 in.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Suitable for Go/NoGo inspection and measuring hole sizes.
  • Includes a certificate of accuracy with NIST traceability.
  • Offers economical wear life for various gaging applications.