Learn More
MSC Vermont Gage Pin Gage, Class ZZ, NoGo, 17.21 mm, Steel
Catalog No. 5000527042
Encompass
This Class ZZ NoGo pin gage, with a 17.21 mm diameter, is constructed from durable tool steel. It is designed for precise 'NoGo' inspection, making it suitable for setting micrometers, verniers, and other measuring devices. The gage features a reversible style and meets Class ZZ tolerance specifications.
  • Class ZZ tolerance of 0.00508 mm
  • NoGo (minus) direction of tolerance
  • Made from durable steel for extended wear life
  • Reversible style for versatile use
  • NIST traceable for accuracy verification
  • 2 inch (50.8 mm) length
  • 10 microinch finish or better
  • Round within 0.0025 mm
  • Marked with size and direction of tolerance
  • Includes a certificate of accuracy
  • Individually packaged for protection
Catalog No. 50-005-27042 Supplier MSC Supplier No. 40444663
This Class ZZ NoGo pin gage, with a 17.21 mm diameter, is constructed from durable tool steel. It is designed for precise 'NoGo' inspection, making it suitable for setting micrometers, verniers, and other measuring devices. The gage features a reversible style and meets Class ZZ tolerance specifications.
  • Class ZZ tolerance of 0.00508 mm
  • NoGo (minus) direction of tolerance
  • Made from durable steel for extended wear life
  • Reversible style for versatile use
  • NIST traceable for accuracy verification
  • 2 inch (50.8 mm) length
  • 10 microinch finish or better
  • Round within 0.0025 mm
  • Marked with size and direction of tolerance
  • Includes a certificate of accuracy
  • Individually packaged for protection