Learn More
MSC Vermont Gage Pin Gage, Class ZZ, NoGo, Steel, 15.50 mm
Catalog No. 5000528519
Encompass
This pin gage is a Class ZZ, NoGo gage made of steel, with a diameter of 15.50 mm. It is designed for setting micrometers, verniers, and other measuring devices for NoGo inspection.
  • Class ZZ
  • Designed for setting micrometers, verniers, and other measuring devices for NoGo inspection
  • 15.5 mm diameter
  • Minus direction of tolerance
  • Steel construction
  • Reversible style
  • Metric system of measurement
  • 0.00508 mm tolerance
Catalog No. 50-005-28519 Supplier MSC Supplier No. 40442956
This pin gage is a Class ZZ, NoGo gage made of steel, with a diameter of 15.50 mm. It is designed for setting micrometers, verniers, and other measuring devices for NoGo inspection.
  • Class ZZ
  • Designed for setting micrometers, verniers, and other measuring devices for NoGo inspection
  • 15.5 mm diameter
  • Minus direction of tolerance
  • Steel construction
  • Reversible style
  • Metric system of measurement
  • 0.00508 mm tolerance