Learn More
MSC VERMONT GAGE Pin Gage, Class ZZ, NoGo, Steel, 18.44 mm
Catalog No. 5000529054
Encompass
This pin gage is designed for NoGo inspection, belonging to Class ZZ and constructed from steel. It features a diameter of 18.44 mm, a reversible style, and operates within the metric system with a tolerance of 0.00508 mm. It is suitable for setting micrometers, verniers, and other measuring devices.
  • Designed for NoGo inspection
  • Class ZZ
  • Made of steel
  • 18.44 mm diameter
  • Reversible style
  • Metric system of measurement
  • Tolerance of 0.00508 mm
  • Minus direction of tolerance
  • Can be used to set micrometers, verniers, and other measuring devices
Catalog No. 50-005-29054 Supplier MSC Supplier No. 40445892
This pin gage is designed for NoGo inspection, belonging to Class ZZ and constructed from steel. It features a diameter of 18.44 mm, a reversible style, and operates within the metric system with a tolerance of 0.00508 mm. It is suitable for setting micrometers, verniers, and other measuring devices.
  • Designed for NoGo inspection
  • Class ZZ
  • Made of steel
  • 18.44 mm diameter
  • Reversible style
  • Metric system of measurement
  • Tolerance of 0.00508 mm
  • Minus direction of tolerance
  • Can be used to set micrometers, verniers, and other measuring devices