Learn More
MSC Vermont Gage Pin Gage, Class ZZ, NoGo, Steel, 19.98 mm
Catalog No. 5000527144
Encompass
This pin gage is a Class ZZ, NoGo type, constructed from steel with a precise diameter of 19.98 mm. It is specifically designed for NoGo inspection and serves as a reliable tool for setting micrometers, verniers, and other measuring devices.
  • Class ZZ
  • NoGo inspection
  • Made from steel
  • 19.98 mm diameter
  • Minus tolerance direction
  • Metric system of measurement
  • Reversible style
  • Used to set micrometers, verniers, and other measuring devices
Catalog No. 50-005-27144 Supplier MSC Supplier No. 40447435
This pin gage is a Class ZZ, NoGo type, constructed from steel with a precise diameter of 19.98 mm. It is specifically designed for NoGo inspection and serves as a reliable tool for setting micrometers, verniers, and other measuring devices.
  • Class ZZ
  • NoGo inspection
  • Made from steel
  • 19.98 mm diameter
  • Minus tolerance direction
  • Metric system of measurement
  • Reversible style
  • Used to set micrometers, verniers, and other measuring devices