Learn More
MSC Vermont Gage Pin Gage, Class ZZ, NoGo, Steel, 3.13 mm
Catalog No. 5000528888
Encompass
This pin gage is a Class ZZ gage, designed for NoGo inspection. It is made of steel and features a 3.13 mm size. This reversible gage utilizes a metric system of measurement with a 0.00508 mm minus tolerance. It is ideal for applications such as setting micrometers, verniers, and other measuring devices.
  • Designed for NoGo inspection
  • Class ZZ gage
  • Reversible style
  • Metric system of measurement
  • 3.13 mm diameter
  • Minus direction of tolerance
  • 0.00508 mm tolerance
  • Ideal for setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-28888 Supplier MSC Supplier No. 40430589
This pin gage is a Class ZZ gage, designed for NoGo inspection. It is made of steel and features a 3.13 mm size. This reversible gage utilizes a metric system of measurement with a 0.00508 mm minus tolerance. It is ideal for applications such as setting micrometers, verniers, and other measuring devices.
  • Designed for NoGo inspection
  • Class ZZ gage
  • Reversible style
  • Metric system of measurement
  • 3.13 mm diameter
  • Minus direction of tolerance
  • 0.00508 mm tolerance
  • Ideal for setting micrometers, verniers, and other measuring devices