Learn More
MSC Vermont Gage Pin Gage, Class ZZ Plus, 0.1675 in Diameter, Tool Steel
Catalog No. 5000510436
Encompass
This pin gage is designed for precise measurement of hole sizes within a tight tolerance. Made from durable tool steel, it offers an economical solution for quick and accurate gaging in various applications.
  • Go (Plus) type, ideal for checking the upper limit of a hole or feature
  • Class ZZ tolerance for high accuracy
  • Constructed from tool steel with a hardness of 60/65 Rc
  • Overall length of 2 in for versatile use
  • Features a surface finish of 10 microinch or better
  • Round within 0.0001 in for consistent measurements
  • Includes a certificate of accuracy with NIST traceability
  • Useful for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations
Catalog No. 50-005-10436 Supplier MSC Supplier No. 88916754
This pin gage is designed for precise measurement of hole sizes within a tight tolerance. Made from durable tool steel, it offers an economical solution for quick and accurate gaging in various applications.
  • Go (Plus) type, ideal for checking the upper limit of a hole or feature
  • Class ZZ tolerance for high accuracy
  • Constructed from tool steel with a hardness of 60/65 Rc
  • Overall length of 2 in for versatile use
  • Features a surface finish of 10 microinch or better
  • Round within 0.0001 in for consistent measurements
  • Includes a certificate of accuracy with NIST traceability
  • Useful for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations