Learn More
MSC Vermont Gage Pin Gage, Class ZZ Plus, 0.4730 in Diameter, 2.0000 in Length, Tool Steel
Catalog No. 5000513795
Encompass
The Vermont Gage Pin Gage, Class ZZ Plus, is a precision measuring tool designed for accurate inspection of hole sizes and for setting other measuring devices. It features a 0.4730 inch diameter and a 2.0000 inch overall length, crafted from durable tool steel.
  • Designed for go inspection and setting various measuring devices.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a 10 microfinish or better for precision.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Tolerance of +.0002/-.0000 in.
Catalog No. 50-005-13795 Supplier MSC Supplier No. 88947304
The Vermont Gage Pin Gage, Class ZZ Plus, is a precision measuring tool designed for accurate inspection of hole sizes and for setting other measuring devices. It features a 0.4730 inch diameter and a 2.0000 inch overall length, crafted from durable tool steel.
  • Designed for go inspection and setting various measuring devices.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a 10 microfinish or better for precision.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Tolerance of +.0002/-.0000 in.