Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Plus, 20.82 mm, Tool Steel
Catalog No. 5000530689
Encompass
Vermont Gage Class ZZ Plus Pin Gages, such as this 20.82 mm diameter pin gage, are economical, general-purpose tools designed for quick and accurate measurements. They are primarily used for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations, making them suitable for various Go/NoGo gaging applications.
  • Class ZZ tolerance of .0002 in (0.005 mm)
  • Go (Plus) tolerance direction
  • Tool steel material; 60/65 Rc hardness
  • 10 microinch finish or better
  • Two in long, round within .0001 in (0.0025 mm)
  • Marked with size and tolerance on gages over .0605 in
  • NIST traceable with certificate of accuracy
  • Offers added wear life
  • Individually packaged
  • Measures hole sizes, distances between holes, and slot widths
  • Supports Go/NoGo gaging and sizing with micrometers
Catalog No. 50-005-30689 Supplier MSC Supplier No. 68111871
Vermont Gage Class ZZ Plus Pin Gages, such as this 20.82 mm diameter pin gage, are economical, general-purpose tools designed for quick and accurate measurements. They are primarily used for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations, making them suitable for various Go/NoGo gaging applications.
  • Class ZZ tolerance of .0002 in (0.005 mm)
  • Go (Plus) tolerance direction
  • Tool steel material; 60/65 Rc hardness
  • 10 microinch finish or better
  • Two in long, round within .0001 in (0.0025 mm)
  • Marked with size and tolerance on gages over .0605 in
  • NIST traceable with certificate of accuracy
  • Offers added wear life
  • Individually packaged
  • Measures hole sizes, distances between holes, and slot widths
  • Supports Go/NoGo gaging and sizing with micrometers