Learn More
MSC Vermont Gage Pin Gage, Class ZZ Plus, 3.84 mm, Tool Steel
Catalog No. 5000512741
Encompass
This pin gage is designed for precise and economical measurement of hole diameters and setting various measuring devices. Crafted from durable heat-treated tool steel, it features a 2-inch length and a Class ZZ Plus tolerance. Each gage includes a Certificate of Accuracy with NIST traceability for reliable measurements.
  • For precise, economical measurement of hole diameters and distances.
  • Features 0.0000/+0.0002-inch plus tolerance.
  • Includes NIST traceability and Certificate of Accuracy.
  • Made from heat-treated tool steel (60-62 RC hardness) with 10 micro-finish.
  • 2-inch long, centerless lapped; corrosion and wear resistant.
  • Electro-etched size for easy identification.
  • Ideal for setting micrometers, calipers, and snap gages.
  • Compatible with GO/NOGO tools for diameter and depth checks.
Catalog No. 50-005-12741 Supplier MSC Supplier No. 68094895
This pin gage is designed for precise and economical measurement of hole diameters and setting various measuring devices. Crafted from durable heat-treated tool steel, it features a 2-inch length and a Class ZZ Plus tolerance. Each gage includes a Certificate of Accuracy with NIST traceability for reliable measurements.
  • For precise, economical measurement of hole diameters and distances.
  • Features 0.0000/+0.0002-inch plus tolerance.
  • Includes NIST traceability and Certificate of Accuracy.
  • Made from heat-treated tool steel (60-62 RC hardness) with 10 micro-finish.
  • 2-inch long, centerless lapped; corrosion and wear resistant.
  • Electro-etched size for easy identification.
  • Ideal for setting micrometers, calipers, and snap gages.
  • Compatible with GO/NOGO tools for diameter and depth checks.