Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Plus Tolerance, 0.3385 in Diameter
Catalog No. 5000510464
Encompass
This precision pin gage is designed for accurately measuring hole sizes within tight specifications. Constructed from durable tool steel, it features a diameter of 0.3385 inch and an overall length of 2 inches. The gage is classified as Class ZZ Plus, offering a tight tolerance of +0.0002/-0.0000 " Its surface boasts a 10 microfinish or better, ensuring reliability and accuracy in various applications. Each gage includes a Certificate of Accuracy with NIST Traceability, confirming its adherence to high measurement standards.
  • Constructed from durable tool steel.
  • Features a hardness of Rc 60/65.
  • Offers a precise tolerance of +0.0002/-0.0000 "
  • Maintains a surface finish of 10 microinch or better.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.
  • Overall length of 2 inches.
Catalog No. 50-005-10464 Supplier MSC Supplier No. 88933858
This precision pin gage is designed for accurately measuring hole sizes within tight specifications. Constructed from durable tool steel, it features a diameter of 0.3385 inch and an overall length of 2 inches. The gage is classified as Class ZZ Plus, offering a tight tolerance of +0.0002/-0.0000 " Its surface boasts a 10 microfinish or better, ensuring reliability and accuracy in various applications. Each gage includes a Certificate of Accuracy with NIST Traceability, confirming its adherence to high measurement standards.
  • Constructed from durable tool steel.
  • Features a hardness of Rc 60/65.
  • Offers a precise tolerance of +0.0002/-0.0000 "
  • Maintains a surface finish of 10 microinch or better.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.
  • Overall length of 2 inches.