Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Plus Tolerance, Steel, 19.50 mm
Catalog No. 5000527342
Encompass
This pin gage is designed for precise measurement with a 19.50 mm size. It features a Class ZZ tolerance and is constructed from durable steel. Its reversible style allows for extended use and metric system compatibility ensures accurate measurements.
  • Class ZZ tolerance for high precision.
  • Plus tolerance for specific measurement needs.
  • Constructed from durable steel for longevity.
  • 19.50 mm size for exact specifications.
  • Reversible style for extended lifespan.
  • Metric system of measurement for broad applicability.
Catalog No. 50-005-27342 Supplier MSC Supplier No. 68110550
This pin gage is designed for precise measurement with a 19.50 mm size. It features a Class ZZ tolerance and is constructed from durable steel. Its reversible style allows for extended use and metric system compatibility ensures accurate measurements.
  • Class ZZ tolerance for high precision.
  • Plus tolerance for specific measurement needs.
  • Constructed from durable steel for longevity.
  • 19.50 mm size for exact specifications.
  • Reversible style for extended lifespan.
  • Metric system of measurement for broad applicability.