Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Steel, 19.99 mm, Minus Tolerance
Catalog No. 5000514654
Encompass
Vermont Gage pin gages are precision measuring devices designed for setting micrometers, verniers, and other measuring equipment, as well as for 'no-go' inspection. These gages are made from steel and feature a reversible style, ensuring durability and ease of use.
  • Class ZZ for precise measurements.
  • Minus tolerance for specific inspection needs.
  • Reversible style for extended tool life.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Ideal for no-go inspection.
  • Metric system of measurement.
  • Diameter of 19.99 mm.
  • Tolerance of 0.00508 mm.
Catalog No. 50-005-14654 Supplier MSC Supplier No. 40447443
Vermont Gage pin gages are precision measuring devices designed for setting micrometers, verniers, and other measuring equipment, as well as for 'no-go' inspection. These gages are made from steel and feature a reversible style, ensuring durability and ease of use.
  • Class ZZ for precise measurements.
  • Minus tolerance for specific inspection needs.
  • Reversible style for extended tool life.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Ideal for no-go inspection.
  • Metric system of measurement.
  • Diameter of 19.99 mm.
  • Tolerance of 0.00508 mm.