Learn More
MSC Vermont Gage Pin Gage, Metric, Class ZZ, Tool Steel
Catalog No. 5000514347
Encompass
These Class ZZ steel gage pins are designed for quick and accurate measurements, ideal for checking hole diameters, distances between holes, slot widths, and locations. They are suitable for Go/NoGo inspection and for setting micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance (0.005 mm).
  • Manufactured from tool steel; 60/65 Rc hardness.
  • 2 inch length.
  • 10 microinch finish or better.
  • Round within 0.0025 mm.
  • Marked with diameter and direction of tolerance on gages over 0.0605 inches.
  • Certificate of accuracy included.
Catalog No. 50-005-14347 Supplier MSC Supplier No. 68107002
These Class ZZ steel gage pins are designed for quick and accurate measurements, ideal for checking hole diameters, distances between holes, slot widths, and locations. They are suitable for Go/NoGo inspection and for setting micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance (0.005 mm).
  • Manufactured from tool steel; 60/65 Rc hardness.
  • 2 inch length.
  • 10 microinch finish or better.
  • Round within 0.0025 mm.
  • Marked with diameter and direction of tolerance on gages over 0.0605 inches.
  • Certificate of accuracy included.