Learn More
MSC Vermont Gage Pin Gage, Minus, 0.0250 in, Class ZZ, Steel
Catalog No. 5000531786
Encompass
This Class ZZ No-Go pin gage, made from durable steel with a black oxide finish, is engineered for precise measurement of hole sizes within a tolerance of +0.0000/-0.0002 inches. With an overall length of 2 inches and a diameter of 0.0250 inches, it is suitable for various industrial and laboratory applications. It includes a Certificate of Accuracy with NIST Traceability and meets ISO 17025 standards, ensuring reliability and precision.
  • Measures hole sizes within +0.0000/-0.0002 in
  • Includes certificate of accuracy with NIST traceability
  • Overall length of 2 in
  • Diameter of 0.0250 in
  • Dimension type is inch
  • Cylindrical shape
  • Finish is black oxide
  • Hardness is RC 60/65
  • Surface finish is 10 microinch or better
  • Meets ISO 17025 standards
Catalog No. 50-005-31786 Supplier MSC Supplier No. 62256177
This Class ZZ No-Go pin gage, made from durable steel with a black oxide finish, is engineered for precise measurement of hole sizes within a tolerance of +0.0000/-0.0002 inches. With an overall length of 2 inches and a diameter of 0.0250 inches, it is suitable for various industrial and laboratory applications. It includes a Certificate of Accuracy with NIST Traceability and meets ISO 17025 standards, ensuring reliability and precision.
  • Measures hole sizes within +0.0000/-0.0002 in
  • Includes certificate of accuracy with NIST traceability
  • Overall length of 2 in
  • Diameter of 0.0250 in
  • Dimension type is inch
  • Cylindrical shape
  • Finish is black oxide
  • Hardness is RC 60/65
  • Surface finish is 10 microinch or better
  • Meets ISO 17025 standards