Learn More
MSC Vermont Gage Pin Gage, Minus, 10.46 mm, Class ZZ, Tool Steel
Catalog No. 5000533960
Encompass
Vermont Gage Class ZZ Minus Pin Gages are an affordable solution for measuring hole sizes, distances between holes, or slot widths. Made from durable tool steel, these gages are heat treated and feature a fine microfinish for accuracy. They are designed for NoGo inspection and come with a certificate of accuracy with NIST traceability.
  • Measures hole diameters, distances between holes, and slot widths
  • Used for no go inspection
  • Made from tool steel
  • Heat treated to 60-65 Rc
  • Features a 10 microinch finish or better
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-33960 Supplier MSC Supplier No. 40437915
Vermont Gage Class ZZ Minus Pin Gages are an affordable solution for measuring hole sizes, distances between holes, or slot widths. Made from durable tool steel, these gages are heat treated and feature a fine microfinish for accuracy. They are designed for NoGo inspection and come with a certificate of accuracy with NIST traceability.
  • Measures hole diameters, distances between holes, and slot widths
  • Used for no go inspection
  • Made from tool steel
  • Heat treated to 60-65 Rc
  • Features a 10 microinch finish or better
  • Includes a certificate of accuracy with NIST traceability