Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.0200 in
Catalog No. 5000537403
Encompass
This pin gage is designed for precise measurement of hole sizes within a tight tolerance of +0.0000/-0.0002 inches. Manufactured from steel with a hardness of RC 60/65, it features a 0.0200 inch diameter and an overall length of 2 inches. This Class ZZ, minus tolerance gage is ideal for quality control applications requiring high accuracy.
  • Cylindrical shape for easy insertion
  • Constructed from durable steel
  • 0.0200 inch nominal diameter
  • Class ZZ tolerance (+0.0000/-0.0002 in) for high precision
  • 2 inch overall length
  • Hardness rated at RC 60/65
  • Surface finish of 10 microinch or better
  • Certificate of accuracy with NIST traceability included
  • Complies with ISO 17025 standards
Catalog No. 50-005-37403 Supplier MSC Supplier No. 62256128
This pin gage is designed for precise measurement of hole sizes within a tight tolerance of +0.0000/-0.0002 inches. Manufactured from steel with a hardness of RC 60/65, it features a 0.0200 inch diameter and an overall length of 2 inches. This Class ZZ, minus tolerance gage is ideal for quality control applications requiring high accuracy.
  • Cylindrical shape for easy insertion
  • Constructed from durable steel
  • 0.0200 inch nominal diameter
  • Class ZZ tolerance (+0.0000/-0.0002 in) for high precision
  • 2 inch overall length
  • Hardness rated at RC 60/65
  • Surface finish of 10 microinch or better
  • Certificate of accuracy with NIST traceability included
  • Complies with ISO 17025 standards