Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.2405 in, Tool Steel
Catalog No. 5000515796
Encompass
This pin gage provides precise measurement for hole sizes, featuring a 0.2405 inch diameter and a Class ZZ designation for high accuracy. It is made from durable tool steel with a Rockwell hardness of 60 to 65 and boasts a superior steel finish (10 microfinish or better). With a 2 inch overall length and a tolerance of +.0000/-.0002, this NoGo (minus) type pin gage is ideal for verifying exact hole dimensions.
  • Durable tool steel construction with Rockwell hardness of 60 to 65.
  • Superior steel finish, 10 microfinish or better.
  • Measures hole sizes with a precise tolerance of +.0000/-.0002 "
  • Includes a certificate of accuracy with NIST traceability.
  • NoGo (minus) type, ideal for verifying minimum hole dimensions.
  • 2 inch overall length.
  • Class ZZ designation for high precision.
Catalog No. 50-005-15796 Supplier MSC Supplier No. 89024053
This pin gage provides precise measurement for hole sizes, featuring a 0.2405 inch diameter and a Class ZZ designation for high accuracy. It is made from durable tool steel with a Rockwell hardness of 60 to 65 and boasts a superior steel finish (10 microfinish or better). With a 2 inch overall length and a tolerance of +.0000/-.0002, this NoGo (minus) type pin gage is ideal for verifying exact hole dimensions.
  • Durable tool steel construction with Rockwell hardness of 60 to 65.
  • Superior steel finish, 10 microfinish or better.
  • Measures hole sizes with a precise tolerance of +.0000/-.0002 "
  • Includes a certificate of accuracy with NIST traceability.
  • NoGo (minus) type, ideal for verifying minimum hole dimensions.
  • 2 inch overall length.
  • Class ZZ designation for high precision.