Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.3335 Inch, Tool Steel
Catalog No. 5000531158
Encompass
This pin gage is constructed from durable tool steel with a 60/65 Rc hardness. It is designed for precise measurement of hole sizes, featuring a 0.3335 inch diameter and a tight tolerance of +.0000/-.0002. This NoGo (Minus) Class ZZ gage ensures accurate checks and comes with a Certificate of Accuracy.
  • Material is tool steel; Rc 60/65
  • Tolerance of +.0000/-.0002
  • Measures hole sizes within +.0000/-.0002
  • Includes certificate of accuracy with NIST traceability
  • NoGo (minus) type for specific gauging
  • Class ZZ for precise measurements
  • Steel finish with 10 microinch finish or better
  • Overall length of 2 in
Catalog No. 50-005-31158 Supplier MSC Supplier No. 89033351
This pin gage is constructed from durable tool steel with a 60/65 Rc hardness. It is designed for precise measurement of hole sizes, featuring a 0.3335 inch diameter and a tight tolerance of +.0000/-.0002. This NoGo (Minus) Class ZZ gage ensures accurate checks and comes with a Certificate of Accuracy.
  • Material is tool steel; Rc 60/65
  • Tolerance of +.0000/-.0002
  • Measures hole sizes within +.0000/-.0002
  • Includes certificate of accuracy with NIST traceability
  • NoGo (minus) type for specific gauging
  • Class ZZ for precise measurements
  • Steel finish with 10 microinch finish or better
  • Overall length of 2 in