Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.5585 Inch, Steel
Catalog No. 5000536678
Encompass
This pin gage is designed for precise measurement applications, featuring a minus tolerance and Class ZZ classification. Made from durable tool steel, it offers accurate sizing for various hole diameters. The imperial system measurement ensures compatibility with common industry standards.
  • Class ZZ for high precision measurements
  • Minus tolerance of +.0000/-.0002
  • Made from robust tool steel with Rc 60/65 hardness
  • Features a 0.5585 inch diameter
  • Overall length of 2 inches
  • Includes a certificate of accuracy with NIST traceability
  • Steel finish with 10 microfinish or better
  • Reversible style for extended use
Catalog No. 50-005-36678 Supplier MSC Supplier No. 89992689
This pin gage is designed for precise measurement applications, featuring a minus tolerance and Class ZZ classification. Made from durable tool steel, it offers accurate sizing for various hole diameters. The imperial system measurement ensures compatibility with common industry standards.
  • Class ZZ for high precision measurements
  • Minus tolerance of +.0000/-.0002
  • Made from robust tool steel with Rc 60/65 hardness
  • Features a 0.5585 inch diameter
  • Overall length of 2 inches
  • Includes a certificate of accuracy with NIST traceability
  • Steel finish with 10 microfinish or better
  • Reversible style for extended use