Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.5590 in, Tool Steel
Catalog No. 5000530713
Encompass
This pin gage is a Class ZZ minus type, featuring a 0.5590 inch diameter. Constructed from durable tool steel, it is designed for precisely measuring hole sizes with a tolerance of +.0000/-.0002 inches.
  • Includes certificate of accuracy with NIST traceability
  • Finish is steel - 10 microfinish or better
  • Overall length is 2 inches
  • Made from tool steel with Rc 60/65 hardness
  • Tolerance of +.0000/-.0002
  • Measures hole sizes within +.0000/-.0002 inches
  • Type is NoGo (Minus)
Catalog No. 50-005-30713 Supplier MSC Supplier No. 89055909
This pin gage is a Class ZZ minus type, featuring a 0.5590 inch diameter. Constructed from durable tool steel, it is designed for precisely measuring hole sizes with a tolerance of +.0000/-.0002 inches.
  • Includes certificate of accuracy with NIST traceability
  • Finish is steel - 10 microfinish or better
  • Overall length is 2 inches
  • Made from tool steel with Rc 60/65 hardness
  • Tolerance of +.0000/-.0002
  • Measures hole sizes within +.0000/-.0002 inches
  • Type is NoGo (Minus)