Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.573 in Diameter, Tool Steel
Catalog No. 5000531902
Encompass
This pin gage, with a diameter of 0.573 in, is designed for precise measurements of hole sizes. It is a NoGo (Minus) type, belonging to Class ZZ, and is constructed from durable tool steel with an Rc 60/65 hardness. The pin gage features a steel finish of 10 microfinish or better, ensuring accuracy. It has a tolerance of +.0000/-.0002 and an overall length of 2 in. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Constructed from durable tool steel with Rc 60/65 hardness.
  • Features a tolerance of +.0000/-.0002.
  • Includes a certificate of accuracy with NIST traceability.
  • Has an overall length of 2 in.
  • Provides a steel finish of 10 microfinish or better.
Catalog No. 50-005-31902 Supplier MSC Supplier No. 89057301
This pin gage, with a diameter of 0.573 in, is designed for precise measurements of hole sizes. It is a NoGo (Minus) type, belonging to Class ZZ, and is constructed from durable tool steel with an Rc 60/65 hardness. The pin gage features a steel finish of 10 microfinish or better, ensuring accuracy. It has a tolerance of +.0000/-.0002 and an overall length of 2 in. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Constructed from durable tool steel with Rc 60/65 hardness.
  • Features a tolerance of +.0000/-.0002.
  • Includes a certificate of accuracy with NIST traceability.
  • Has an overall length of 2 in.
  • Provides a steel finish of 10 microfinish or better.