Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.7170 in Diameter, Tool Steel
Catalog No. 5000531469
Encompass
This pin gage is designed for precise measurement of hole sizes within a specified tolerance. Constructed from durable tool steel, it features a 0.7170 diameter, a Class ZZ designation, and a minus type configuration for accurate gaging applications. Each gage is supplied with a certificate of accuracy.
  • Measures hole sizes within +.0000/-.0002.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a steel finish of 10 microfinish or better.
  • Includes a certificate of accuracy with NIST traceability.
  • Has an overall length of 2 in.
  • Designed as a class ZZ, nogo (minus) type.
Catalog No. 50-005-31469 Supplier MSC Supplier No. 89071708
This pin gage is designed for precise measurement of hole sizes within a specified tolerance. Constructed from durable tool steel, it features a 0.7170 diameter, a Class ZZ designation, and a minus type configuration for accurate gaging applications. Each gage is supplied with a certificate of accuracy.
  • Measures hole sizes within +.0000/-.0002.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a steel finish of 10 microfinish or better.
  • Includes a certificate of accuracy with NIST traceability.
  • Has an overall length of 2 in.
  • Designed as a class ZZ, nogo (minus) type.