Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.7770, Tool Steel
Catalog No. 5000511430
Encompass
This Class ZZ Minus Pin Gage, made from tool steel, has a diameter of 0.7770 inches and is designed to measure hole sizes within a tolerance of +.0000/-.0002 inches.
  • Includes certificate of accuracy with NIST traceability.
  • Manufactured from tool steel with Rc 60/65 hardness.
  • Features a steel finish of 10 microfinish or better.
  • Has an overall length of 2 inches.
  • Designed for measuring hole sizes with high precision.
Catalog No. 50-005-11430 Supplier MSC Supplier No. 89077705
This Class ZZ Minus Pin Gage, made from tool steel, has a diameter of 0.7770 inches and is designed to measure hole sizes within a tolerance of +.0000/-.0002 inches.
  • Includes certificate of accuracy with NIST traceability.
  • Manufactured from tool steel with Rc 60/65 hardness.
  • Features a steel finish of 10 microfinish or better.
  • Has an overall length of 2 inches.
  • Designed for measuring hole sizes with high precision.