Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.8880 in Diameter
Catalog No. 5000531385
Encompass
This Vermont Gage Pin Gage is a NoGo (Minus) type, Class ZZ, designed to accurately measure hole sizes. It features a 0.8880 in diameter and is constructed from durable tool steel with a Rc 60/65 hardness, ensuring precision and longevity.
  • Measures hole sizes within +.0000/-.0002 in tolerance.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a 10 microfinish or better for smooth surface.
  • Includes a certificate of accuracy with NIST traceability.
  • Overall length of 2 in.
Catalog No. 50-005-31385 Supplier MSC Supplier No. 89088801
This Vermont Gage Pin Gage is a NoGo (Minus) type, Class ZZ, designed to accurately measure hole sizes. It features a 0.8880 in diameter and is constructed from durable tool steel with a Rc 60/65 hardness, ensuring precision and longevity.
  • Measures hole sizes within +.0000/-.0002 in tolerance.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a 10 microfinish or better for smooth surface.
  • Includes a certificate of accuracy with NIST traceability.
  • Overall length of 2 in.