Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 14.04mm, Tool Steel
Catalog No. 5000534051
Encompass
This pin gage is designed for precise measurement of hole sizes. Made from durable tool steel, it features a 14.04mm diameter and a Class ZZ tolerance. Its overall length is 2 inches, and it has a fine steel finish. This NoGo (minus) type gage is suitable for various industrial applications.
  • Measures hole sizes within +.000/-.005"
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from tool steel with Rc 60/65 hardness
  • Features a steel finish of 10 microfinish or better
  • Designed with an overall length of 2 inches
  • Conforms to Class ZZ tolerance standards
Catalog No. 50-005-34051 Supplier MSC Supplier No. 40441495
This pin gage is designed for precise measurement of hole sizes. Made from durable tool steel, it features a 14.04mm diameter and a Class ZZ tolerance. Its overall length is 2 inches, and it has a fine steel finish. This NoGo (minus) type gage is suitable for various industrial applications.
  • Measures hole sizes within +.000/-.005"
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from tool steel with Rc 60/65 hardness
  • Features a steel finish of 10 microfinish or better
  • Designed with an overall length of 2 inches
  • Conforms to Class ZZ tolerance standards